2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053111-500MG
053111-2.5G
101902-332EA
364.62
USD
101902-332
101902-334
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-methylbutanamide hydrochloride
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₂₁ClN₂O₂ |
MDL Number:
MFCD13562583 CAS Number: 1236262-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...