3-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053110-500MG 053110-2.5G
101902-328EA 364.62 USD
101902-328 101902-330
3-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride
3-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562582
CAS Number: 1220028-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR