N-(2-Hydroxy-1,1-dimethylethyl)-2-(methylamino)acetamide hydrochloride
Supplier: Matrix Scientific
053109-500MG
053109-2.5G
101902-324EA
364.62
USD
101902-324
101902-326
N-(2-Hydroxy-1,1-dimethylethyl)-2-(methylamino)acetamide hydrochloride
N-(2-Hydroxy-1,1-dimethylethyl)-2-(methylamino)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₇ClN₂O₂ MW: 196.68 g/mol |
MDL Number:
MFCD13562581 CAS Number: 1219957-60-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...