2-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053108-500MG 053108-2.5G
101902-320EA 364.62 USD
101902-320 101902-322
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562580
CAS Number: 1236256-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR