2-Amino-N-(2-hydroxy-1,1-dimethylethyl)acetamide hydrochloride

Supplier: Matrix Scientific

053107-500MG 053107-2.5G
101902-316EA 364.62 USD
101902-316 101902-318
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)acetamide hydrochloride
2-Amino-N-(2-hydroxy-1,1-dimethylethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD13562579
CAS Number: 1220035-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR