2-[(6-Chloro-2-pyridinyl)amino]-2-methyl-1-propanol
Supplier: Matrix Scientific
053106-500MG
053106-2.5G
101902-312EA
364.62
USD
101902-312
101902-314
2-[(6-Chloro-2-pyridinyl)amino]-2-methyl-1-propanol
2-[(6-Chloro-2-pyridinyl)amino]-2-methyl-1-propanol
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O |
MDL Number:
MFCD13562578 CAS Number: 1219963-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...