2-[(5-Bromo-2-pyridinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053105-500MG 053105-2.5G
101902-308EA 364.62 USD
101902-308 101902-310
2-[(5-Bromo-2-pyridinyl)amino]-2-methyl-1-propanol
2-[(5-Bromo-2-pyridinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD12184825
CAS Number: 1154894-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR