2-[(3-Amino-2-pyridinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053102-500MG 053102-2.5G
101902-296EA 364.62 USD
101902-296 101902-298
2-[(3-Amino-2-pyridinyl)amino]-2-methyl-1-propanol
2-[(3-Amino-2-pyridinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₉H₁₅N₃O
MDL Number: MFCD12184030
CAS Number: 1155070-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR