2-[(2-Hydroxy-1,1-dimethylethyl)amino]nicotinic acid

Supplier: Matrix Scientific

053101-500MG 053101-2.5G
101902-292EA 364.62 USD
101902-292 101902-294
2-[(2-Hydroxy-1,1-dimethylethyl)amino]nicotinic acid
2-[(2-Hydroxy-1,1-dimethylethyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD13562576
CAS Number: 1220035-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR