2-[(2-Hydroxy-1,1-dimethylethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053100-500MG
053100-2.5G
101902-288EA
364.62
USD
101902-288
101902-290
2-[(2-Hydroxy-1,1-dimethylethyl)amino]isonicotinic acid
2-[(2-Hydroxy-1,1-dimethylethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₄N₂O₃ |
MDL Number:
MFCD13562575 CAS Number: 1220035-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...