2-[(6-Chloro-2-pyrazinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053099-500MG 053099-2.5G
101902-284EA 364.62 USD
101902-284 101902-286
2-[(6-Chloro-2-pyrazinyl)amino]-2-methyl-1-propanol
2-[(6-Chloro-2-pyrazinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MDL Number: MFCD13562574
CAS Number: 1220038-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR