2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-methyl-1-propanol
Supplier: Matrix Scientific
053098-500MG
053098-2.5G
101902-280EA
364.62
USD
101902-280
101902-282
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-methyl-1-propanol
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}-2-methyl-1-propanol
Hazard: Irritant
Formula:
C₁₀H₁₂ClF₃N₂O |
MDL Number:
MFCD12416178 CAS Number: 1157109-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...