2-[(6-Chloro-4-pyrimidinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053097-500MG 053097-2.5G
101902-276EA 364.62 USD
101902-276 101902-278
2-[(6-Chloro-4-pyrimidinyl)amino]-2-methyl-1-propanol
2-[(6-Chloro-4-pyrimidinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MDL Number: MFCD13562573
CAS Number: 1219963-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR