2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-methyl-1-propanol

Supplier: Matrix Scientific

053096-500MG 053096-2.5G
101902-272EA 364.62 USD
101902-272 101902-274
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-methyl-1-propanol
2-[(5-Bromo-4-methyl-2-pyridinyl)amino]-2-methyl-1-propanol

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD13562572
CAS Number: 1220030-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR