4-[(2-Hydroxy-1,1-dimethylethyl)amino]-3-nitrobenzoic acid
Supplier: Matrix Scientific
053095-500MG
053095-2.5G
101902-268EA
364.62
USD
101902-268
101902-270
4-[(2-Hydroxy-1,1-dimethylethyl)amino]-3-nitrobenzoic acid
4-[(2-Hydroxy-1,1-dimethylethyl)amino]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₅ |
MDL Number:
MFCD13562571 CAS Number: 1220030-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...