Ethyl-3-amino-4-[(2-hydroxy-1,1-dimethylethyl)amino]benzoate

Supplier: Matrix Scientific

053094-500MG 053094-2.5G
101902-264EA 364.62 USD
101902-264 101902-266
Ethyl-3-amino-4-[(2-hydroxy-1,1-dimethylethyl)amino]benzoate
Ethyl-3-amino-4-[(2-hydroxy-1,1-dimethylethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃
MDL Number: MFCD13562570
CAS Number: 1220038-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR