3-Amino-4-chloro-N-[2-(dimethylamino)ethyl]benzenesulfonamide

Supplier: Matrix Scientific

053093-500MG 053093-2.5G
101902-260EA 364.62 USD
101902-260 101902-262
3-Amino-4-chloro-N-[2-(dimethylamino)ethyl]benzenesulfonamide
3-Amino-4-chloro-N-[2-(dimethylamino)ethyl]benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₆ClN₃O₂S
MDL Number: MFCD11125124
CAS Number: 1040026-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR