N-[2-(Dimethylamino)ethyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride

Supplier: Matrix Scientific

053092-500MG 053092-2.5G
101902-256EA 364.62 USD
101902-256 101902-258
N-[2-(Dimethylamino)ethyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁Cl₂N₅O
MDL Number: MFCD13562569
CAS Number: 1220035-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR