N-[2-(Dimethylamino)ethyl]-4-piperidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053091-500MG 053091-2.5G
101902-252EA 364.62 USD
101902-252 101902-254
N-[2-(Dimethylamino)ethyl]-4-piperidinecarboxamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-4-piperidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃O
MDL Number: MFCD10686831
CAS Number: 886504-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR