N-[2-(Dimethylamino)ethyl]-3-piperidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053090-500MG 053090-2.5G
101902-248EA 364.62 USD
101902-248 101902-250
N-[2-(Dimethylamino)ethyl]-3-piperidinecarboxamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-3-piperidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃O
MDL Number: MFCD13562568
CAS Number: 1220038-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR