N-[2-(Dimethylamino)ethyl]-2-piperidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053089-500MG 053089-2.5G
101902-244EA 364.62 USD
101902-244 101902-246
N-[2-(Dimethylamino)ethyl]-2-piperidinecarboxamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-2-piperidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₃Cl₂N₃O
MW: 272.22 g/mol
MDL Number: MFCD13562567
CAS Number: 1236263-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR