N-[2-(Dimethylamino)ethyl]-2-pyrrolidinecarboxamide dihydrochloride

Supplier: Matrix Scientific

053088-500MG 053088-2.5G
101902-240EA 364.62 USD
101902-240 101902-242
N-[2-(Dimethylamino)ethyl]-2-pyrrolidinecarboxamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-2-pyrrolidinecarboxamide dihydrochloride

Hazard: Irritant

Formula: C₉H₂₁Cl₂N₃O
MDL Number: MFCD13562566
CAS Number: 1236256-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR