2-Amino-N-[2-(dimethylamino)ethyl]-3-phenylpropanamide dihydrochloride

Supplier: Matrix Scientific

053087-500MG 053087-2.5G
101902-236EA 364.62 USD
101902-236 101902-238
2-Amino-N-[2-(dimethylamino)ethyl]-3-phenylpropanamide dihydrochloride
2-Amino-N-[2-(dimethylamino)ethyl]-3-phenylpropanamide dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₃Cl₂N₃O
MW: 308.25 g/mol
MDL Number: MFCD13562565
CAS Number: 1246172-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR