2-Amino-N-[2-(dimethylamino)ethyl]-3-methylbutanamide dihydrochloride

Supplier: Matrix Scientific

053085-500MG 053085-2.5G
101902-228EA 364.62 USD
101902-228 101902-230
2-Amino-N-[2-(dimethylamino)ethyl]-3-methylbutanamide dihydrochloride
2-Amino-N-[2-(dimethylamino)ethyl]-3-methylbutanamide dihydrochloride

Hazard: Irritant

Formula: C₉H₂₃Cl₂N₃O
MDL Number: MFCD13562563
CAS Number: 1236261-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR