3-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride

Supplier: Matrix Scientific

053084-500MG 053084-2.5G
101902-224EA 384.62 USD
101902-224 101902-226
3-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride
3-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride

Hazard: Irritant

Formula: C₇H₁₉Cl₂N₃O
MDL Number: MFCD13562562
CAS Number: 1144082-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR