N-[2-(Dimethylamino)ethyl]-2-(methylamino)acetamide dihydrochloride
Supplier: Matrix Scientific
053083-500MG
053083-2.5G
101902-220EA
364.62
USD
101902-220
101902-222
N-[2-(Dimethylamino)ethyl]-2-(methylamino)acetamide dihydrochloride
N-[2-(Dimethylamino)ethyl]-2-(methylamino)acetamide dihydrochloride
Hazard: Irritant
Formula:
C₇H₁₉Cl₂N₃O MW: 232.15 g/mol |
MDL Number:
MFCD13562561 CAS Number: 1220037-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...