2-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride

Supplier: Matrix Scientific

053082-500MG 053082-2.5G
101902-216EA 364.62 USD
101902-216 101902-218
2-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride
2-Amino-N-[2-(dimethylamino)ethyl]propanamide dihydrochloride

Hazard: Irritant

Formula: C₇H₁₉Cl₂N₃O
MDL Number: MFCD13562560
CAS Number: 1246172-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR