2-Amino-N-[2-(dimethylamino)ethyl]acetamide dihydrochloride
Supplier: Matrix Scientific
053081-500MG
053081-2.5G
101902-212EA
364.62
USD
101902-212
101902-214
2-Amino-N-[2-(dimethylamino)ethyl]acetamide dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₇Cl₂N₃O |
MDL Number:
MFCD13562559 CAS Number: 1144082-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...