N1-(6-Chloro-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Supplier: Matrix Scientific
053080-500MG
053080-2.5G
101902-208EA
364.62
USD
101902-208
101902-210
N1-(6-Chloro-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(6-Chloro-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Hazard: Irritant
Formula:
C₉H₁₄ClN₃ |
MDL Number:
MFCD13562558 CAS Number: 3298-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...