N1-(5-Bromo-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Supplier: Matrix Scientific
053079-500MG
053079-2.5G
101902-204EA
364.62
USD
101902-204
101902-206
N1-(5-Bromo-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(5-Bromo-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Hazard: Irritant
Formula:
C₉H₁₄BrN₃ |
MDL Number:
MFCD11125892 CAS Number: 112357-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...