N1-(5-Bromo-3-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine

Supplier: Matrix Scientific

053078-500MG 053078-2.5G
101902-200EA 364.62 USD
101902-200 101902-202
N1-(5-Bromo-3-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(5-Bromo-3-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine

Hazard: Irritant

Formula: C₁₀H₁₆BrN₃
MDL Number: MFCD13562557
CAS Number: 1220035-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR