2-Chloro-N-[2-(dimethylamino)ethyl]nicotinamide
Supplier: Matrix Scientific
053077-500MG
053077-2.5G
101902-196EA
364.62
USD
101902-196
101902-198
2-Chloro-N-[2-(dimethylamino)ethyl]nicotinamide
2-Chloro-N-[2-(dimethylamino)ethyl]nicotinamide
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃O |
MDL Number:
MFCD09803546 CAS Number: 1016674-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...