N2-[2-(Dimethylamino)ethyl]-2,3-pyridinediamine

Supplier: Matrix Scientific

053076-500MG 053076-2.5G
101902-192EA 384.62 USD
101902-192 101902-194
N2-[2-(Dimethylamino)ethyl]-2,3-pyridinediamine
N2-[2-(Dimethylamino)ethyl]-2,3-pyridinediamine

Hazard: Irritant

Formula: C₉H₁₆N₄
MDL Number: MFCD11186951
CAS Number: 834798-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR