2-{[2-(Dimethylamino)ethyl]amino}nicotinic acid
Supplier: Matrix Scientific
053075-500MG
053075-2.5G
101902-188EA
364.62
USD
101902-188
101902-190
2-{[2-(Dimethylamino)ethyl]amino}nicotinic acid
2-{[2-(Dimethylamino)ethyl]amino}nicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O₂ |
MDL Number:
MFCD11132026 CAS Number: 115891-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...