2-{[2-(Dimethylamino)ethyl]amino}isonicotinic acid

Supplier: Matrix Scientific

053074-500MG 053074-2.5G
101902-184EA 364.62 USD
101902-184 101902-186
2-{[2-(Dimethylamino)ethyl]amino}isonicotinic acid
2-{[2-(Dimethylamino)ethyl]amino}isonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MDL Number: MFCD11132023
CAS Number: 1019371-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR