N1-(6-Chloro-2-pyrazinyl)-N2,N2-dimethyl-1,2-ethanediamine

Supplier: Matrix Scientific

053073-500MG 053073-2.5G
101902-180EA 384.62 USD
101902-180 101902-182
N1-(6-Chloro-2-pyrazinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(6-Chloro-2-pyrazinyl)-N2,N2-dimethyl-1,2-ethanediamine

Hazard: Irritant

Formula: C₈H₁₃ClN₄
MDL Number: MFCD13562556
CAS Number: 1219957-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR