N1-(6-Chloro-4-pyrimidinyl)-N2,N2-dimethyl-1,2-ethanediamine

Supplier: Matrix Scientific

053071-500MG 053071-2.5G
101902-172EA 364.62 USD
101902-172 101902-174
N1-(6-Chloro-4-pyrimidinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(6-Chloro-4-pyrimidinyl)-N2,N2-dimethyl-1,2-ethanediamine

Hazard: Irritant

Formula: C₈H₁₃ClN₄
MDL Number: MFCD13562555
CAS Number: 1220028-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR