N1-(5-Bromo-4-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Supplier: Matrix Scientific
053070-500MG
053070-2.5G
101902-168EA
149.23
USD
101902-168
101902-170
N1-(5-Bromo-4-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
N1-(5-Bromo-4-methyl-2-pyridinyl)-N2,N2-dimethyl-1,2-ethanediamine
Hazard: Irritant
Formula:
C₁₀H₁₆BrN₃ |
MDL Number:
MFCD13562554 CAS Number: 1220037-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...