4-{[2-(Dimethylamino)ethyl]amino}-3-nitrobenzoic acid
Supplier: Matrix Scientific
053069-500MG
053069-2.5G
101902-164EA
364.62
USD
101902-164
101902-166
4-{[2-(Dimethylamino)ethyl]amino}-3-nitrobenzoic acid
4-{[2-(Dimethylamino)ethyl]amino}-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₁H₁₅N₃O₄ |
MDL Number:
MFCD11523528 CAS Number: 104144-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...