Ethyl-3-amino-4-{[2-(dimethylamino)ethyl]amino}benzoate

Supplier: Matrix Scientific

053068-500MG 053068-2.5G
101902-160EA 364.62 USD
101902-160 101902-162
Ethyl-3-amino-4-{[2-(dimethylamino)ethyl]amino}benzoate
Ethyl-3-amino-4-{[2-(dimethylamino)ethyl]amino}benzoate

Hazard: Irritant

Formula: C₁₃H₂₁N₃O₂
MDL Number: MFCD13343660
CAS Number: 1220035-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR