N-Cyclopentyl-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053064-500MG
053064-2.5G
101902-144EA
364.62
USD
101902-144
101902-146
N-Cyclopentyl-3-piperidinecarboxamide hydrochloride
N-Cyclopentyl-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O MW: 232.76 g/mol |
MDL Number:
MFCD13562551 CAS Number: 937724-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...