2-Amino-N-cyclopentyl-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053061-500MG 053061-2.5G
101902-132EA 364.62 USD
101902-132 101902-134
2-Amino-N-cyclopentyl-3-phenylpropanamide hydrochloride
2-Amino-N-cyclopentyl-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MDL Number: MFCD13562548
CAS Number: 1246172-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR