2-Amino-N-cyclopentyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

053059-500MG 053059-2.5G
101902-124EA 364.62 USD
101902-124 101902-126
2-Amino-N-cyclopentyl-3-methylbutanamide hydrochloride
2-Amino-N-cyclopentyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD13562546
CAS Number: 1236262-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR