N-Cyclopentyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

053057-500MG 053057-2.5G
101902-116EA 364.62 USD
101902-116 101902-118
N-Cyclopentyl-2-(methylamino)acetamide hydrochloride
N-Cyclopentyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MW: 192.69 g/mol
MDL Number: MFCD13562544
CAS Number: 1220028-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR