2-Amino-N-cyclopentylacetamide hydrochloride

Supplier: Matrix Scientific

053055-500MG 053055-2.5G
101902-108EA 364.62 USD
101902-108 101902-110
2-Amino-N-cyclopentylacetamide hydrochloride
2-Amino-N-cyclopentylacetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD08752575
CAS Number: 1187931-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR