5-Bromo-N-cyclopentyl-2-pyridinamine
Supplier: Matrix Scientific
053053-500MG
053053-2.5G
101902-100EA
149.23
USD
101902-100
101902-102
5-Bromo-N-cyclopentyl-2-pyridinamine
5-Bromo-N-cyclopentyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂ |
MDL Number:
MFCD11118485 CAS Number: 1036468-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...