5-Bromo-N-cyclopentyl-3-methyl-2-pyridinamine
Supplier: Matrix Scientific
053052-500MG
053052-2.5G
101902-096EA
364.62
USD
101902-096
101902-098
5-Bromo-N-cyclopentyl-3-methyl-2-pyridinamine
5-Bromo-N-cyclopentyl-3-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂ |
MDL Number:
MFCD13562542 CAS Number: 1219957-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...