N2-Cyclopentyl-2,3-pyridinediamine

Supplier: Matrix Scientific

053050-500MG 053050-2.5G
101902-088EA 166.15 USD
101902-088 101902-090
N2-Cyclopentyl-2,3-pyridinediamine
N2-Cyclopentyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
MDL Number: MFCD08692158
CAS Number: 951523-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR