2-(Cyclopentylamino)isonicotinic acid
Supplier: Matrix Scientific
053048-500MG
053048-2.5G
101902-080EA
364.62
USD
101902-080
101902-082
2-(Cyclopentylamino)isonicotinic acid
2-(Cyclopentylamino)isonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ |
MDL Number:
MFCD11132015 CAS Number: 1019127-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...