6-Chloro-N-cyclopentyl-2-pyrazinamine
Supplier: Matrix Scientific
053047-500MG
053047-2.5G
101902-076EA
364.62
USD
101902-076
101902-078
6-Chloro-N-cyclopentyl-2-pyrazinamine
6-Chloro-N-cyclopentyl-2-pyrazinamine
Hazard: Irritant
Formula:
C₉H₁₂ClN₃ |
MDL Number:
MFCD10597764 CAS Number: 642459-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...